Assay Details
Target Gene Details
Entrez Gene ID: | 92291 |
Gene Name: | calpain 13 |
Gene Aliases: |
- |
Location: |
Chr.2:30610382-30809527 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPN13 | XM_017005265.1 | XP_016860754.1 |
Target Gene Details
Entrez Gene ID: | 253558 |
Gene Name: | lysocardiolipin acyltransferase 1 |
Gene Aliases: |
1AGPAT8, AGPAT8, ALCAT1, HSRG1849, LYCAT, UNQ1849 |
Location: |
Chr.2:30447226-30644225 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCLAT1 | NM_001002257.2 | NP_001002257.1 | ||
| NM_001304445.1 | NP_001291374.1 | |||
| NM_001304446.1 | NP_001291375.1 | |||
| NM_182551.4 | NP_872357.2 | |||
| XM_005264244.1 | XP_005264301.1 | |||
| XM_005264245.3 | XP_005264302.1 | |||
| XM_011532741.2 | XP_011531043.1 | |||
| XM_011532742.2 | XP_011531044.1 | |||
| XM_011532743.2 | XP_011531045.1 | |||
| XM_011532744.2 | XP_011531046.1 | |||
| XM_017003746.1 | XP_016859235.1 | |||
| AK296965.1 | ||||
| AK297262.1 | ||||
| AK302562.1 | ||||
| AK311294.1 | ||||
| AY358702.1 | AAQ89065.1 | |||
| BC146817.1 | ||||
| BX647953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3590201 | Chr.2:30589923 - 30627405 on Build GRCh38 | Loss |
|
| esv2762445 | Chr.2:30592132 - 30626477 on Build GRCh38 | Loss |
|
| nsv1003432 | Chr.2:30592120 - 30626477 on Build GRCh38 | Loss |
|
| dgv689e199 | Chr.2:30589882 - 30627418 on Build GRCh38 | Deletion |
|
| esv3583706 | Chr.2:30592119 - 30626734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map