Assay Details
Target Gene Details
Entrez Gene ID: | 80034 |
Gene Name: | cysteine and serine rich nuclear protein 3 |
Gene Aliases: |
FAM130A2, PPP1R73, TAIP-2, TAIP2 |
Location: |
Chr.2:165469647-165689407 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSRNP3 | NM_001172173.1 | NP_001165644.1 | ||
| AK126129.1 | BAC86454.1 |
More Information
Set Membership: |
|

Genomic Map