Assay Details
Target Gene Details
Entrez Gene ID: | 386597 |
Gene Name: | RNF144A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:6912276-6918682 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF144A-AS1 | NR_033997.1 | |||
| AK057135.1 | 2 | 2021 | ||
| AK090737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833326 | Chr.2:6844896 - 7002433 on Build GRCh38 | Loss |
|

Genomic Map