Assay Details
Target Gene Details
Entrez Gene ID: | 3344 |
Gene Name: | forkhead box N2 |
Gene Aliases: |
HTLF |
Location: |
Chr.2:48314640-48379295 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXN2 | NM_002158.3 | NP_002149.2 | ||
| XM_005264282.2 | XP_005264339.1 | |||
| XM_005264283.2 | XP_005264340.1 | |||
| XM_006712001.3 | XP_006712064.1 | |||
| XM_006712002.3 | XP_006712065.1 | |||
| XM_011532808.2 | XP_011531110.1 | |||
| XM_017003956.1 | XP_016859445.1 | |||
| XM_017003957.1 | XP_016859446.1 | |||
| XM_017003958.1 | XP_016859447.1 | |||
| XM_017003959.1 | XP_016859448.1 | |||
| BC063305.1 | AAH63305.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762252 | Chr.2:48327011 - 48513954 on Build GRCh38 | Gain |
|
| nsv519831 | Chr.2:47845716 - 48338892 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map