Assay Details
Target Gene Details
Entrez Gene ID: | 23040 |
Gene Name: | myelin transcription factor 1 like |
Gene Aliases: |
MRD39, NZF1, ZC2H2C2, ZC2HC4B |
Location: |
Chr.2:1789113-2331388 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Gene Details
Entrez Gene ID: | 730811 |
Gene Name: | MYT1L antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:2319232-2327108 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYT1L-AS1 | NR_024468.1 | |||
| AK057398.1 | ||||
| AK126008.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833237 | Chr.2:2170069 - 2350137 on Build GRCh38 | Loss |
|
| nsv1001231 | Chr.2:1961134 - 2365066 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map