Assay Details
Target Gene Details
Entrez Gene ID: | 10057 |
Gene Name: | ATP binding cassette subfamily C member 5 |
Gene Aliases: |
ABC33, EST277145, MOAT-C, MOATC, MRP5, SMRP, pABC11 |
Location: |
Chr.3:183919934-184018010 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC5 | NM_001023587.2 | NP_001018881.1 | ||
| NM_001320032.1 | NP_001306961.1 | |||
| NM_005688.3 | NP_005679.2 | |||
| NR_135125.1 | ||||
| XM_005247058.4 | XP_005247115.1 | |||
| XM_005247059.4 | XP_005247116.1 | |||
| XM_011512314.1 | XP_011510616.1 | |||
| XM_011512315.1 | XP_011510617.1 | |||
| XM_017005493.1 | XP_016860982.1 | |||
| XM_017005494.1 | XP_016860983.1 | |||
| AB019002.1 | BAA76608.1 | |||
| AB209454.1 | BAD92691.1 | |||
| AF104942.1 | AAD04169.1 | |||
| AF146074.1 | AAD37716.1 | |||
| AL524917.3 | ||||
| AY196484.1 | AAO49801.1 | |||
| AY754874.1 | AAW82948.1 | |||
| AY754875.1 | AAW82949.1 | |||
| AY754876.1 | AAW82950.1 | |||
| BC015334.1 | AAH15334.1 | |||
| BC050744.1 | AAH50744.1 | |||
| BC140771.1 | ||||
| BC142670.1 | ||||
| BC142719.1 | ||||
| U83661.2 | AAB71758.2 |
Target Gene Details
Entrez Gene ID: | 100873982 |
Gene Name: | ABCC5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:184006338-184011419 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC5-AS1 | NR_046570.1 | |||
| BE549568.1 |
More Information
Set Membership: |
|

Genomic Map