Assay Details
Target Gene Details
Entrez Gene ID: | 84561 |
Gene Name: | solute carrier family 12 member 8 |
Gene Aliases: |
CCC9 |
Location: |
Chr.3:125082636-125212765 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC12A8 | NM_001195483.1 | NP_001182412.1 | ||
| NM_024628.5 | NP_078904.3 | |||
| AF345197.1 | AAO49174.1 | |||
| AF390442.1 | AAM73657.1 | |||
| AK026841.1 | BAB15571.1 | |||
| AK123001.1 | ||||
| AK308484.1 | ||||
| BC020506.1 | AAH20506.1 | |||
| BC063528.1 | AAH63528.1 | |||
| BC126158.1 | ||||
| BC126160.1 | ||||
| BX538023.1 | CAD97969.1 | |||
| CR457363.1 | CAG33644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv990342 | Chr.3:125084201 - 125084785 on Build GRCh38 | Gain |
|
| nsv523278 | Chr.3:125084044 - 125084947 on Build GRCh38 | Loss |
|
| nsv3982 | Chr.3:125078149 - 125106568 on Build GRCh38 | Insertion |
|
| esv34126 | Chr.3:125045570 - 125149965 on Build GRCh38 | Loss |
|
| nsv10311 | Chr.3:125073408 - 125703762 on Build GRCh38 | Gain |
|
| nsv1001677 | Chr.3:124015843 - 125108694 on Build GRCh38 | Loss |
|
| nsv1160974 | Chr.3:124577493 - 125300995 on Build GRCh38 | Duplication |
|
| nsv829712 | Chr.3:125038838 - 125183948 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map