Assay Details
Target Gene Details
Entrez Gene ID: | 51319 |
Gene Name: | arginine and serine rich coiled-coil 1 |
Gene Aliases: |
BM-011, SFRS21, SRrp53 |
Location: |
Chr.3:158110052-158544835 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSRC1 | NM_001271834.1 | NP_001258763.1 | ||
| NM_001271838.1 | NP_001258767.1 | |||
| NM_016625.3 | NP_057709.2 | |||
| AF208853.1 | AAF64267.1 | |||
| AK290334.1 | ||||
| AK300388.1 | ||||
| BC006982.1 | AAH06982.1 | |||
| BC010357.1 | AAH10357.1 | |||
| DA273191.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829768 | Chr.3:158115089 - 158294281 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map