Assay Details
Target Gene Details
Entrez Gene ID: | 64084 |
Gene Name: | calsyntenin 2 |
Gene Aliases: |
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma |
Location: |
Chr.3:139935185-140577397 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLSTN2 | NM_022131.2 | NP_071414.2 | ||
| AJ278018.1 | CAC14924.1 | |||
| AK096432.1 | ||||
| AK315308.1 | ||||
| AY753303.1 | AAV30553.1 | |||
| BC103496.1 | AAI03497.1 | |||
| BC103506.1 | AAI03507.1 | |||
| BC104485.1 | AAI04486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597943 | Chr.3:139941861 - 139946951 on Build GRCh38 | Loss |
|
| nsv998423 | Chr.3:139807604 - 140228494 on Build GRCh38 | Gain |
|
| nsv591859 | Chr.3:139943096 - 139985332 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73867231] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map