Assay Details
Target Gene Details
Entrez Gene ID: | 1232 |
Gene Name: | C-C motif chemokine receptor 3 |
Gene Aliases: |
CC-CKR-3, CD193, CKR3, CMKBR3 |
Location: |
Chr.3:46210699-46266706 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR3 | NM_001164680.1 | NP_001158152.1 | ||
| NM_001837.3 | NP_001828.1 | |||
| NM_178328.1 | NP_847898.1 | |||
| NM_178329.2 | NP_847899.1 | |||
| XM_006712960.3 | XP_006713023.1 | |||
| XM_011533335.2 | XP_011531637.1 | |||
| XM_017005685.1 | XP_016861174.1 | |||
| XM_017005686.1 | XP_016861175.1 | |||
| AF224495.1 | AAK49027.1 | |||
| AF262299.1 | AAL85628.1 | |||
| AF262300.1 | AAL85629.1 | |||
| AF262301.1 | AAL85630.1 | |||
| AF262302.1 | AAL85631.1 | |||
| AF262303.1 | AAL85632.1 | |||
| AF262304.1 | ||||
| BC110297.1 | AAI10298.1 | |||
| BC130318.1 | ||||
| BC130320.1 |
More Information
Set Membership: |
|

Genomic Map