Assay Details
Target Gene Details
Entrez Gene ID: | 54872 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class G |
Gene Aliases: |
GPI7, LAS21, MRT53, PRO4405, RLGS1930 |
Location: |
Chr.4:499200-539921 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599372 | Chr.4:473707 - 505788 on Build GRCh38 | Loss |
|
| nsv1001899 | Chr.4:68917 - 523947 on Build GRCh38 | Loss |
|
| nsv1006781 | Chr.4:343385 - 560798 on Build GRCh38 | Gain |
|
| nsv593204 | Chr.4:476314 - 534500 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map