Assay Details
Target Gene Details
| Entrez Gene ID: | 2868 | 
| Gene Name: | G protein-coupled receptor kinase 4 | 
| Gene Aliases: | GPRK2L, GPRK4, GRK4a, IT11 | 
| Location: | Chr.4:2963504-3040760 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3575673 | Chr.4:2957494 - 3022996 on Build GRCh38 | Gain |  NOP14  GRK4 | 
| dgv5082n100 | Chr.4:2956087 - 3017315 on Build GRCh38 | Gain |  NOP14  GRK4 | 
| nsv593438 | Chr.4:2946729 - 3028370 on Build GRCh38 | Gain |  NOP14-AS1  NOP14  GRK4 | 
| esv3599415 | Chr.4:2958013 - 3019896 on Build GRCh38 | Gain |  NOP14  GRK4 | 
| esv3599412 | Chr.4:2940424 - 3030117 on Build GRCh38 | Gain |  NOP14-AS1  NOP14  GRK4 | 
| nsv593437 | Chr.4:2904558 - 3233253 on Build GRCh38 | Gain |  NOP14-AS1  HTT-AS  HTT  ADD1  MFSD10  NOP14  GRK4 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top