Assay Details
Target Gene Details
Entrez Gene ID: | 4886 |
Gene Name: | neuropeptide Y receptor Y1 |
Gene Aliases: |
NPY1-R, NPYR |
Location: |
Chr.4:163323962-163344847 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPY1R | NM_000909.5 | NP_000900.1 | ||
| XM_005263031.3 | XP_005263088.1 | |||
| XM_011532010.2 | XP_011530312.1 | |||
| AK296624.1 | ||||
| AK298282.1 | ||||
| AK312578.1 | ||||
| BC036657.2 | AAH36657.1 | |||
| BC071720.1 | AAH71720.1 | |||
| DA531980.1 | ||||
| M88461.1 | AAA73215.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015505 | Chr.4:161989825 - 163516137 on Build GRCh38 | Gain |
|
| nsv1030184 | Chr.4:162402427 - 164455529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map