Assay Details
Target Gene Details
Entrez Gene ID: | 306 |
Gene Name: | annexin A3 |
Gene Aliases: |
ANX3 |
Location: |
Chr.4:78551588-78610451 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANXA3 | NM_005139.2 | NP_005130.1 | ||
| AK313945.1 | ||||
| BC000871.1 | AAH00871.1 | |||
| BX336217.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006026 | Chr.4:78111837 - 79966019 on Build GRCh38 | Loss |
|
| nsv1009633 | Chr.4:77966419 - 78731082 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map