Assay Details
Target Gene Details
Entrez Gene ID: | 59350 |
Gene Name: | relaxin/insulin like family peptide receptor 1 |
Gene Aliases: |
LGR7, RXFPR1 |
Location: |
Chr.4:158521711-158653372 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602833 | Chr.4:158437503 - 158589628 on Build GRCh38 | Gain |
|
| nsv830124 | Chr.4:158518665 - 158643187 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map