Assay Details
Target Gene Details
Entrez Gene ID: | 10941 |
Gene Name: | UDP glucuronosyltransferase family 2 member A1 complex locus |
Gene Aliases: |
UDPGT2A1 |
Location: |
Chr.4:69588417-69653249 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT2A1 | NM_001252274.2 | NP_001239203.2 | ||
| NM_001252275.2 | NP_001239204.2 | |||
| NM_001301239.1 | NP_001288168.1 | |||
| NM_006798.4 | NP_006789.3 | |||
| AJ006054.1 | CAB41974.1 | |||
| AK304249.1 | ||||
| AK314209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3600919 | Chr.4:69621467 - 69651053 on Build GRCh38 | Loss |
|
| dgv5318n100 | Chr.4:69619810 - 69656037 on Build GRCh38 | Loss |
|
| dgv659n27 | Chr.4:69622642 - 69656037 on Build GRCh38 | Loss |
|
| esv3569511 | Chr.4:69621459 - 69650802 on Build GRCh38 | Loss |
|
| nsv594616 | Chr.4:69596324 - 69681535 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map