Assay Details
Target Gene Details
Entrez Gene ID: | 83891 |
Gene Name: | sorting nexin 25 |
Gene Aliases: |
MSTP043, SBBI31 |
Location: |
Chr.4:185204237-185363971 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX25 | NM_001317781.1 | NP_001304710.1 | ||
| XM_005263267.4 | XP_005263324.1 | |||
| XM_011532302.2 | XP_011530604.1 | |||
| XM_011532303.2 | XP_011530605.1 | |||
| XM_011532304.2 | XP_011530606.1 | |||
| XM_011532305.2 | XP_011530607.1 | |||
| XM_011532306.2 | XP_011530608.1 | |||
| XM_011532307.2 | XP_011530609.1 | |||
| XM_011532308.2 | XP_011530610.1 | |||
| XM_017008680.1 | XP_016864169.1 | |||
| XM_017008681.1 | XP_016864170.1 | |||
| XM_017008682.1 | XP_016864171.1 | |||
| XM_017008684.1 | XP_016864173.1 | |||
| XM_017008686.1 | XP_016864175.1 | |||
| XM_017008687.1 | XP_016864176.1 | |||
| AW968034.1 | ||||
| AY601647.1 | AAT98627.1 | |||
| BM554003.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461859 | Chr.4:184907523 - 185314394 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72706074] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map