Assay Details
Target Gene Details
Entrez Gene ID: | 441024 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like |
Gene Aliases: |
- |
Location: |
Chr.4:74123280-74303099 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569519 | Chr.4:74142954 - 74255926 on Build GRCh38 | Loss |
|
| nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain |
|
| nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain |
|
| dgv9134n54 | Chr.4:74147724 - 74240768 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map