Assay Details
Target Gene Details
Entrez Gene ID: | 550112 |
Gene Name: | UBA6 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:67701278-67722505 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA6-AS1 | NR_015439.1 | 2 | 297 | |
| AK093682.1 | 2 | 214 | ||
| AK129519.1 | 2 | 242 | ||
| DA770515.1 | 2 | 297 |

Genomic Map