Assay Details
Target Gene Details
Entrez Gene ID: | 132430 |
Gene Name: | poly(A) binding protein cytoplasmic 4 like |
Gene Aliases: |
- |
Location: |
Chr.4:133948459-134201748 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PABPC4L |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv9274n54 | Chr.4:133948569 - 134063406 on Build GRCh38 | Loss |
|
| nsv1023249 | Chr.4:133946159 - 134064530 on Build GRCh38 | Loss |
|
| dgv5396n100 | Chr.4:133889406 - 134023164 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115372996] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map