Assay Details
Target Gene Details
Entrez Gene ID: | 441024 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like |
Gene Aliases: |
- |
Location: |
Chr.4:74123280-74303099 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTHFD2L | XM_017008225.1 | XP_016863714.1 | ||
| XM_017008226.1 | XP_016863715.1 | |||
| XM_017008231.1 | XP_016863720.1 | |||
| XM_017008232.1 | XP_016863721.1 | |||
| BC032771.1 | AAH32771.1 | |||
| BG566009.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain |
|
| nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72652946] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map