Assay Details
Target Gene Details
Entrez Gene ID: | 4887 |
Gene Name: | neuropeptide Y receptor Y2 |
Gene Aliases: |
NPY2-R |
Location: |
Chr.4:155173514-155217078 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPY2R | XM_005263033.4 | XP_005263090.1 |
More Information
Set Membership: |
|

Genomic Map