Assay Details
Target Gene Details
Entrez Gene ID: | 4086 |
Gene Name: | SMAD family member 1 |
Gene Aliases: |
BSP-1, BSP1, JV4-1, JV41, MADH1, MADR1 |
Location: |
Chr.4:145481306-145559176 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD1 | NM_005900.2 | NP_005891.1 | ||
| XM_006714217.2 | XP_006714280.1 | |||
| XM_011531961.1 | XP_011530263.1 | |||
| BC001878.1 | AAH01878.1 | |||
| BQ962660.1 | ||||
| BU163870.1 | ||||
| U59423.1 | AAB06852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1074222 | Chr.4:145482047 - 145482448 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map