Assay Details
Target Gene Details
Entrez Gene ID: | 345062 |
Gene Name: | protease, serine 48 |
Gene Aliases: |
ESSPL |
Location: |
Chr.4:151276482-151291856 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS48 | XM_011531922.2 | 1 | 352 | XP_011530224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028814 | Chr.4:150874356 - 152285445 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map