Assay Details
Target Gene Details
Entrez Gene ID: | 8001 |
Gene Name: | glycine receptor alpha 3 |
Gene Aliases: |
- |
Location: |
Chr.4:174642011-174829568 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLRA3 | NM_001042543.2 | NP_001036008.1 | ||
| NM_006529.3 | NP_006520.2 | |||
| XM_011532268.2 | XP_011530570.1 | |||
| XM_017008630.1 | XP_016864119.1 | |||
| AF018157.1 | AAC39918.1 | |||
| BC036086.1 | AAH36086.1 | |||
| U93917.1 | AAC39917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3603233 | Chr.4:174643234 - 174652749 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78378188] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map