Assay Details
Target Gene Details
Entrez Gene ID: | 641365 |
Gene Name: | long intergenic non-protein coding RNA 616 |
Gene Aliases: |
- |
Location: |
Chr.4:138027422-138130685 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00616 | NR_037866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830080 | Chr.4:138031664 - 138223572 on Build GRCh38 | Gain |
|
| nsv1033022 | Chr.4:137999933 - 138081308 on Build GRCh38 | Loss |
|
| nsv526874 | Chr.4:138056019 - 138073250 on Build GRCh38 | Loss |
|

Genomic Map