Assay Details
Target Gene Details
Entrez Gene ID: | 4086 |
Gene Name: | SMAD family member 1 |
Gene Aliases: |
BSP-1, BSP1, JV4-1, JV41, MADH1, MADR1 |
Location: |
Chr.4:145481306-145559176 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD1 | NM_001003688.1 | NP_001003688.1 | ||
| NM_005900.2 | NP_005891.1 | |||
| XM_005262992.3 | XP_005263049.1 | |||
| XM_006714217.2 | XP_006714280.1 | |||
| XM_011531961.1 | XP_011530263.1 | |||
| XM_011531962.1 | XP_011530264.1 | |||
| XM_011531963.1 | XP_011530265.1 | |||
| XM_011531964.1 | XP_011530266.1 | |||
| AK293055.1 | ||||
| BC001878.1 | AAH01878.1 | |||
| BQ962660.1 | ||||
| BU163870.1 | ||||
| U54826.1 | AAC50493.1 | |||
| U59423.1 | AAB06852.1 | |||
| U59912.1 | AAC50790.1 |
Target Gene Details
Entrez Gene ID: | 101927659 |
Gene Name: | SMAD1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.4:145497049-145502980 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD1-AS2 | NR_108077.1 | |||
| BX098661.1 | ||||
| HY049088.1 |
More Information
Set Membership: |
|

Genomic Map