Assay Details
Target Gene Details
Entrez Gene ID: | 2668 |
Gene Name: | glial cell derived neurotrophic factor |
Gene Aliases: |
ATF1, ATF2, HFB1-GDNF, HSCR3 |
Location: |
Chr.5:37812677-37840044 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDNF | XM_017009337.1 | 1 | 111 | XP_016864826.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv597850 | Chr.5:37839624 - 37840141 on Build GRCh38 | Gain |
|
| esv3604748 | Chr.5:37720912 - 37848930 on Build GRCh38 | Gain |
|
| dgv1593e212 | Chr.5:37832461 - 37848988 on Build GRCh38 | Loss |
|
| nsv597849 | Chr.5:37839516 - 37840540 on Build GRCh38 | Gain |
|
| dgv9728n54 | Chr.5:37839675 - 37840407 on Build GRCh38 | Loss |
|
| esv26026 | Chr.5:37839534 - 37840654 on Build GRCh38 | Loss |
|
| nsv597854 | Chr.5:37839952 - 37840540 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map