Assay Details
Target Gene Details
Entrez Gene ID: | 8622 |
Gene Name: | phosphodiesterase 8B |
Gene Aliases: |
ADSD, PPNAD3 |
Location: |
Chr.5:77180252-77428256 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE8B | XM_005248623.4 | XP_005248680.1 | ||
| XM_005248624.4 | XP_005248681.1 | |||
| XM_011543701.2 | XP_011542003.1 | |||
| XM_017010007.1 | XP_016865496.1 | |||
| XM_017010008.1 | XP_016865497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469515 | Chr.5:77080181 - 77230847 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117818206] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map