Assay Details
Target Gene Details
Entrez Gene ID: | 80762 |
Gene Name: | Nedd4 family interacting protein 1 |
Gene Aliases: |
N4WBP5 |
Location: |
Chr.5:142108759-142154443 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDFIP1 | NM_030571.3 | NP_085048.1 | ||
| AB097010.1 | BAC77363.1 | |||
| AB097037.1 | BAC77390.1 | |||
| AI929639.1 | ||||
| AK075495.1 | BAC11652.1 | |||
| AK075524.1 | BAC11670.1 | |||
| AK315481.1 | ||||
| AY192728.1 | AAP13460.1 | |||
| BC004317.1 | AAH04317.1 | |||
| DA236494.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1154058 | Chr.5:141632251 - 142199943 on Build GRCh38 | Deletion |
|
| esv3606993 | Chr.5:142115399 - 142117367 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map