Assay Details
Target Gene Details
Entrez Gene ID: | 105379045 |
Gene Name: | uncharacterized LOC105379045 |
Gene Aliases: |
- |
Location: |
Chr.5:77769539-77776195 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379045 | XM_011543774.2 | 1 | 6313 | XP_011542076.1 |
Target Gene Details
Entrez Gene ID: | 6902 |
Gene Name: | tubulin folding cofactor A |
Gene Aliases: |
- |
Location: |
Chr.5:77691169-77776361 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCA | NM_001297738.1 | NP_001284667.1 | ||
| NM_001297740.1 | NP_001284669.1 | |||
| NM_004607.2 | NP_004598.1 | |||
| AF038952.1 | AAC39866.1 | |||
| AK300031.1 | ||||
| AK312207.1 | ||||
| BC018210.1 | AAH18210.1 | |||
| BC105285.1 | AAI05286.1 | |||
| BT007354.1 | AAP36018.1 | |||
| CR542170.1 | CAG46967.1 | |||
| CR542186.1 | CAG46983.1 | |||
| DC312139.1 | ||||
| DW436723.1 | ||||
| HY015414.1 |
More Information
Set Membership: |
|

Genomic Map