Assay Details
Target Gene Details
Entrez Gene ID: | 3550 |
Gene Name: | IK cytokine, down-regulator of HLA II |
Gene Aliases: |
CSA2, RED, RER |
Location: |
Chr.5:140647799-140662480 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IK | NM_006083.3 | NP_006074.2 | ||
| AF182645.1 | AAD55447.1 | |||
| AJ005579.1 | CAA06607.1 | |||
| AK225924.1 | ||||
| AK293626.1 | ||||
| AK308730.1 | ||||
| AK312363.1 | ||||
| AL050296.1 | CAB43395.2 | |||
| AY007094.1 | AAG01985.1 | |||
| BC013005.2 | AAH13005.1 | |||
| BC071964.1 | AAH71964.1 |
More Information
Set Membership: |
|

Genomic Map