Assay Details
Target Gene Details
Entrez Gene ID: | 4552 |
Gene Name: | 5-methyltetrahydrofolate-homocysteine methyltransferase reductase |
Gene Aliases: |
MSR, cblE |
Location: |
Chr.5:7869104-7901124 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTRR | NM_002454.2 | NP_002445.2 | ||
| NM_024010.2 | NP_076915.2 | |||
| NR_134480.1 | ||||
| NR_134481.1 | ||||
| NR_134482.1 | ||||
| AF025794.1 | AAC39667.1 | |||
| AF121214.1 | AAF16876.1 | |||
| BC054816.1 | ||||
| BC109216.1 | AAI09217.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508345 | Chr.5:7890779 - 7933358 on Build GRCh38 | Deletion |
|
| nsv597005 | Chr.5:7860291 - 8092964 on Build GRCh38 | Gain |
|
| nsv597003 | Chr.5:7538124 - 8706231 on Build GRCh38 | Gain |
|
| dgv334n21 | Chr.5:7886996 - 7896398 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map