Assay Details
Target Gene Details
Entrez Gene ID: | 153657 |
Gene Name: | tetratricopeptide repeat domain 23 like |
Gene Aliases: |
MC25-1 |
Location: |
Chr.5:34839164-34900511 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC23L | NM_001317949.1 | NP_001304878.1 | ||
| NM_144725.3 | NP_653326.3 | |||
| XM_011513984.2 | XP_011512286.1 | |||
| XM_011513985.2 | XP_011512287.1 | |||
| XM_011513987.2 | XP_011512289.1 | |||
| XM_017009119.1 | XP_016864608.1 | |||
| XM_017009120.1 | XP_016864609.1 | |||
| XM_017009121.1 | XP_016864610.1 | |||
| XM_017009122.1 | XP_016864611.1 | |||
| XM_017009123.1 | XP_016864612.1 | |||
| XM_017009124.1 | XP_016864613.1 | |||
| XM_017009125.1 | XP_016864614.1 | |||
| XM_017009126.1 | XP_016864615.1 | |||
| AK058168.1 | BAB71699.1 | |||
| AK293829.1 | ||||
| AY498865.1 | AAR92152.1 | |||
| JN799997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604684 | Chr.5:34661786 - 34882578 on Build GRCh38 | Gain |
|
| nsv522144 | Chr.5:34778266 - 34898199 on Build GRCh38 | Loss |
|
| esv3604679 | Chr.5:34492621 - 34936206 on Build GRCh38 | Gain |
|
| dgv5629n100 | Chr.5:34798496 - 34983041 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map