Assay Details
Target Gene Details
Entrez Gene ID: | 91937 |
Gene Name: | T-cell immunoglobulin and mucin domain containing 4 |
Gene Aliases: |
SMUCKLER, TIM4 |
Location: |
Chr.5:156915750-156963255 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIMD4 | NM_001146726.1 | NP_001140198.1 | ||
| NM_138379.2 | NP_612388.2 | |||
| XM_017010021.1 | XP_016865510.1 | |||
| AK131025.1 | ||||
| AK301337.1 | ||||
| AK312746.1 | ||||
| BC008988.1 | AAH08988.1 | |||
| JX049980.1 | ||||
| KU178832.1 | ||||
| KU178833.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607302 | Chr.5:156839545 - 157055054 on Build GRCh38 | Gain |
|
| esv2730953 | Chr.5:156931614 - 156932634 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map