Assay Details
Target Gene Details
Entrez Gene ID: | 102546226 |
Gene Name: | uncharacterized LOC102546226 |
Gene Aliases: |
- |
Location: |
Chr.5:88408982-88439090 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102546226 | NR_105021.1 |
Target Gene Details
Entrez Gene ID: | 100505894 |
Gene Name: | TMEM161B antisense RNA 1 |
Gene Aliases: |
linc-POLR3G-8 |
Location: |
Chr.5:88268882-88436674 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM161B-AS1 | NR_039993.1 | |||
| NR_039994.2 | ||||
| NR_039995.1 | ||||
| NR_105019.1 | ||||
| NR_105020.1 | ||||
| BF672668.1 | ||||
| BM719684.1 | ||||
| CN260565.1 | ||||
| DA317999.1 |

Genomic Map