Assay Details
Target Gene Details
Entrez Gene ID: | 2668 |
Gene Name: | glial cell derived neurotrophic factor |
Gene Aliases: |
ATF1, ATF2, HFB1-GDNF, HSCR3 |
Location: |
Chr.5:37812677-37840044 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDNF | NM_000514.3 | NP_000505.1 | ||
| NM_001190468.1 | NP_001177397.1 | |||
| NM_001190469.1 | NP_001177398.1 | |||
| NM_001278098.1 | NP_001265027.1 | |||
| NM_199231.2 | NP_954701.1 | |||
| XM_011514030.2 | XP_011512332.1 | |||
| XM_017009337.1 | XP_016864826.1 | |||
| AF053748.1 | AAD43139.1 | |||
| AJ001897.1 | CAA05075.1 | |||
| AJ001898.1 | CAA05076.1 | |||
| AJ001899.1 | CAA05077.1 | |||
| AJ001900.1 | CAA05078.1 | |||
| AY052832.1 | AAL11017.1 | |||
| BC069119.1 | AAH69119.1 | |||
| BC069369.1 | AAH69369.1 | |||
| BC128108.1 | ||||
| BC128109.1 | ||||
| CR541923.1 | CAG46721.1 | |||
| DQ235474.1 | ||||
| KU178093.1 | ||||
| KU178094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604748 | Chr.5:37720912 - 37848930 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map