Assay Details
Target Gene Details
Entrez Gene ID: | 814 |
Gene Name: | calcium/calmodulin dependent protein kinase IV |
Gene Aliases: |
CaMK IV, CaMK-GR, caMK |
Location: |
Chr.5:111223653-111498503 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK4 | NM_001323374.1 | NP_001310303.1 | ||
| NM_001323375.1 | NP_001310304.1 | |||
| NM_001323376.1 | NP_001310305.1 | |||
| NM_001323377.1 | NP_001310306.1 | |||
| NM_001744.5 | NP_001735.1 | |||
| AK311662.1 | ||||
| AK312840.1 | ||||
| BC016695.1 | AAH16695.2 | |||
| BC025687.1 | AAH25687.1 | |||
| BG721313.1 | ||||
| D30742.1 | BAA06403.1 | |||
| DA114495.1 | ||||
| DA396146.1 | ||||
| L17000.1 | AAA35639.1 | |||
| L24959.1 | AAA18251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3606259 | Chr.5:110985051 - 111262481 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114227090] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map