Assay Details
Target Gene Details
Entrez Gene ID: | 3842 |
Gene Name: | transportin 1 |
Gene Aliases: |
IPO2, KPNB2, MIP, MIP1, TRN |
Location: |
Chr.5:72816591-72914388 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNPO1 | NM_002270.3 | NP_002261.3 | ||
| XM_005248501.1 | XP_005248558.1 | |||
| AK299671.1 | ||||
| AK301021.1 | ||||
| AK308177.1 | ||||
| BC040340.1 | AAH40340.1 | |||
| U70322.1 | AAC50723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950089 | Chr.5:72809774 - 72848273 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map