Assay Details
Target Gene Details
Entrez Gene ID: | 81545 |
Gene Name: | F-box protein 38 |
Gene Aliases: |
Fbx38, HMN2D, MOKA, SP329 |
Location: |
Chr.5:148383935-148442836 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO38 | NM_001271723.1 | NP_001258652.1 | ||
| NM_030793.4 | NP_110420.3 | |||
| XM_005268513.1 | XP_005268570.1 | |||
| XM_006714797.1 | XP_006714860.1 | |||
| XM_017009899.1 | XP_016865388.1 | |||
| AK024024.1 | BAB14783.1 | |||
| AK308414.1 | ||||
| BC033454.1 | AAH33454.1 | |||
| BI459318.1 | ||||
| DA399611.1 |
More Information
Set Membership: |
|

Genomic Map