Assay Details
Target Gene Details
Entrez Gene ID: | 79888 |
Gene Name: | lysophosphatidylcholine acyltransferase 1 |
Gene Aliases: |
AGPAT10, AGPAT9, AYTL2, PFAAP3, lpcat |
Location: |
Chr.5:1461427-1523961 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPCAT1 | NM_024830.3 | NP_079106.3 | ||
| XM_005248373.3 | XP_005248430.1 | |||
| XM_011514132.1 | XP_011512434.1 | |||
| XM_011514134.1 | XP_011512436.1 | |||
| XM_017009879.1 | XP_016865368.1 | |||
| AB244719.2 | ||||
| AF530061.1 | AAQ09945.1 | |||
| AK022499.1 | BAB14061.1 | |||
| AK022505.1 | BAB14065.1 | |||
| AK090444.1 | BAC03425.1 | |||
| AK095552.1 | ||||
| AL831864.1 | CAD38556.1 | |||
| BC020166.1 | AAH20166.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461910 | Chr.5:1423790 - 1468172 on Build GRCh38 | Loss |
|
| nsv596629 | Chr.5:457146 - 1468172 on Build GRCh38 | Gain |
|
| nsv830187 | Chr.5:1317991 - 1509711 on Build GRCh38 | Loss |
|
| dgv9536n54 | Chr.5:1411297 - 1468172 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map