Assay Details
Target Gene Details
Entrez Gene ID: | 30820 |
Gene Name: | potassium voltage-gated channel interacting protein 1 |
Gene Aliases: |
KCHIP1, VABP |
Location: |
Chr.5:170353055-170736632 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNIP1 | NM_001034837.2 | NP_001030009.1 | ||
| NM_001034838.2 | NP_001030010.1 | |||
| NM_001278339.1 | NP_001265268.1 | |||
| NM_001278340.1 | NP_001265269.1 | |||
| NM_014592.3 | NP_055407.1 | |||
| XM_017009407.1 | XP_016864896.1 | |||
| AF199597.1 | AAF33682.1 | |||
| AI884645.1 | ||||
| AK074390.1 | ||||
| AK301775.1 | ||||
| AY170821.1 | AAN77491.1 | |||
| AY780424.1 | AAV51968.1 | |||
| BC050375.1 | AAH50375.1 | |||
| BF514334.1 | ||||
| DC420716.1 | ||||
| DQ148476.1 | AAZ77793.1 | |||
| DQ148477.1 | AAZ77794.1 | |||
| DQ148478.1 | AAZ77795.1 | |||
| DQ148479.1 | AAZ77796.1 |
Target Gene Details
Entrez Gene ID: | 105377716 |
Gene Name: | uncharacterized LOC105377716 |
Gene Aliases: |
- |
Location: |
Chr.5:170639158-170681470 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105377716 | NR_136214.1 | |||
| AA626712.1 | ||||
| BX116959.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600262 | Chr.5:170661487 - 171342848 on Build GRCh38 | Gain |
|
| nsv830549 | Chr.5:170569400 - 170724385 on Build GRCh38 | Gain |
|
| nsv5128 | Chr.5:170666368 - 170700845 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75804525] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map