Assay Details
Target Gene Details
Entrez Gene ID: | 133688 |
Gene Name: | UDP glycosyltransferase family 3 member A1 |
Gene Aliases: |
- |
Location: |
Chr.5:35953089-36001055 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT3A1 | NM_152404.3 | NP_689617.3 | ||
| XM_005248243.4 | XP_005248300.1 | |||
| XM_011513957.2 | XP_011512259.1 | |||
| XM_011513958.2 | XP_011512260.1 | |||
| XM_011513959.2 | XP_011512261.1 | |||
| XM_011513961.2 | XP_011512263.1 | |||
| AK057066.1 | BAB71358.1 | |||
| AK091977.1 | ||||
| AK125803.1 | ||||
| AK290809.1 | ||||
| AK296136.1 | ||||
| AK303770.1 | ||||
| AK308046.1 | ||||
| AK315851.1 | ||||
| BC068446.1 | AAH68446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604707 | Chr.5:35513484 - 36501574 on Build GRCh38 | Gain |
|
| nsv830261 | Chr.5:35862982 - 36017946 on Build GRCh38 | Gain |
|
| nsv471011 | Chr.5:35860966 - 36003115 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map