Assay Details
Target Gene Details
Entrez Gene ID: | 101926905 |
Gene Name: | uncharacterized LOC101926905 |
Gene Aliases: |
- |
Location: |
Chr.5:141046205-141078031 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101926905 | NR_105056.2 | |||
| BG193185.1 | ||||
| HY003594.1 | ||||
| HY010779.1 | ||||
| HY018974.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
| nsv428127 | Chr.5:141016768 - 141319173 on Build GRCh38 | Loss |
|

Genomic Map