Assay Details
Target Gene Details
Entrez Gene ID: | 54557 |
Gene Name: | small glutamine rich tetratricopeptide repeat containing beta |
Gene Aliases: |
SGT2 |
Location: |
Chr.5:65665928-65722821 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGTB | NM_019072.2 | NP_061945.1 | ||
| XM_005248548.3 | XP_005248605.1 | |||
| XM_017009597.1 | XP_016865086.1 | |||
| AF075050.1 | AAC28459.1 | |||
| AF368281.1 | AAP29459.1 | |||
| AK096321.1 | BAC04761.1 | |||
| BC012044.1 | AAH12044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894204 | Chr.5:65586109 - 65714937 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76591726] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map