Assay Details
Target Gene Details
Entrez Gene ID: | 643226 |
Gene Name: | glutaredoxin and cysteine rich domain containing 2 |
Gene Aliases: |
DFNB101 |
Location: |
Chr.5:145858538-145931673 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRXCR2 | XM_017009708.1 | XP_016865197.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76774026] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map