Assay Details
Target Gene Details
Entrez Gene ID: | 153201 |
Gene Name: | solute carrier family 36 member 2 |
Gene Aliases: |
PAT2, TRAMD1 |
Location: |
Chr.5:151314978-151347590 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC36A2 | NM_181776.2 | NP_861441.2 | ||
| XM_005268377.3 | XP_005268434.1 | |||
| XM_006714756.3 | XP_006714819.1 | |||
| XM_017009084.1 | XP_016864573.1 | |||
| AK309575.1 | ||||
| AY162214.1 | AAO11788.1 | |||
| AY228364.1 | AAO74832.1 | |||
| BC101100.1 | AAI01101.1 | |||
| BC101101.1 | AAI01102.1 | |||
| BC101102.1 | AAI01103.1 | |||
| BC101103.2 | AAI01104.1 |
More Information
Set Membership: |
|

Genomic Map