Assay Details
Target Gene Details
Entrez Gene ID: | 23107 |
Gene Name: | mitochondrial ribosomal protein S27 |
Gene Aliases: |
MRP-S27, S27mt |
Location: |
Chr.5:72219409-72320257 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS27 | NM_001286748.1 | NP_001273677.1 | ||
| NM_001286751.1 | NP_001273680.1 | |||
| NM_015084.2 | NP_055899.2 | |||
| AK025417.1 | ||||
| AK299412.1 | ||||
| AK300111.1 | ||||
| AK309065.1 | ||||
| BC064902.1 | AAH64902.1 | |||
| D87453.1 | ||||
| DC341760.1 |
More Information
Set Membership: |
|

Genomic Map