Assay Details
Target Gene Details
Entrez Gene ID: | 404203 |
Gene Name: | serine peptidase inhibitor, Kazal type 6 |
Gene Aliases: |
BUSI2, UNQ844 |
Location: |
Chr.5:148202794-148215137 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINK6 | NM_001195290.1 | NP_001182219.1 | ||
| NM_205841.3 | NP_995313.2 | |||
| AY358716.1 | AAQ89078.1 | |||
| BC032003.1 | AAH32003.1 | |||
| BE122852.1 | ||||
| GQ504704.1 | ||||
| GQ504705.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607101 | Chr.5:148020480 - 148213612 on Build GRCh38 | Loss |
|
| esv3607102 | Chr.5:148026584 - 148359140 on Build GRCh38 | Loss |
|
| nsv830515 | Chr.5:148081289 - 148222684 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114269639] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map