Assay Details
Target Gene Details
Entrez Gene ID: | 6507 |
Gene Name: | solute carrier family 1 member 3 |
Gene Aliases: |
EA6, EAAT1, GLAST, GLAST1 |
Location: |
Chr.5:36606355-36688334 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC1A3 | NM_001166695.2 | NP_001160167.1 | ||
| NM_001289939.1 | NP_001276868.1 | |||
| NM_001289940.1 | NP_001276869.1 | |||
| NM_004172.4 | NP_004163.3 | |||
| XM_005248342.2 | XP_005248399.1 | |||
| XM_011514084.2 | XP_011512386.1 | |||
| AK057823.1 | ||||
| AK293886.1 | ||||
| AK295032.1 | ||||
| AK295754.1 | ||||
| AK312304.1 | ||||
| AY954110.1 | AAY28724.1 | |||
| BC037310.2 | AAH37310.1 | |||
| D26443.1 | BAA05462.1 | |||
| L19158.1 | ||||
| U03504.1 | AAA50428.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762516 | Chr.5:36599283 - 36613099 on Build GRCh38 | Loss |
|
| nsv830263 | Chr.5:36576402 - 36754991 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75882017] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map